Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppard | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013141.2 | NP_037273.2 | 8 | 1553 | ||
XM_006256166.2 | XP_006256228.1 | 8 | 2365 | |||
XM_006256167.2 | XP_006256229.1 | 7 | 2472 | |||
XM_006256168.2 | XP_006256230.1 | 7 | 3367 | |||
XM_008772797.1 | XP_008771019.1 | 7 | 2600 | |||
GenBank mRNA | CK356918.1 | - | 1 | 134 | ||
U40064.1 | - | 8 | 1574 | |||
U75918.1 | - | 8 | 1567 |