Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013144.1 | NP_037276.1 | 3 | 738 | ||
GenBank mRNA | BC078889.1 | - | 3 | 720 | 7129185 | |
M58634.1 | - | 3 | 738 | |||
M89791.1 | - | 3 | 699 |