Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134445.1 | NP_001127917.1 | 6, 7 | 942 | ||
NM_012137.3 | NP_036269.1 | 5, 6 | 910 | |||
XM_005270707.2 | XP_005270764.1 | 5, 6 | 619 | |||
XM_005270710.2 | XP_005270767.1 | 5, 6 | 838 | |||
XM_011541158.1 | XP_011539460.1 | 7, 8 | 942 | |||
XM_017000889.1 | XP_016856378.1 | 5, 6 | 596 | |||
XM_017000890.1 | XP_016856379.1 | 5, 6 | 2348 | |||
GenBank mRNA | AK294752.1 | - | 5, 6 | 535 | ||
AK302531.1 | - | 5, 6 | 563 | |||
AK304876.1 | - | 6, 7 | 946 | |||
AK309225.1 | - | 5, 6 | 519 | |||
AK312432.1 | - | 5, 6 | 871 | |||
BC033680.1 | - | 5, 6 | 848 | 5189970 | ||
BC043235.2 | - | 5, 6 | 632 | |||
BX648145.1 | - | 6, 7 | 829 | |||
EU794605.1 | - | 5, 6 | 848 |