Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013156.2 | NP_037288.1 | 3 | 291 | ||
GenBank mRNA | AB630323.1 | - | 2 | 232 | ||
BC063175.1 | - | 3 | 306 | 6918389 | ||
FQ209613.1 | - | 3 | 302 | |||
FQ218649.1 | - | 3 | 300 | |||
FQ228725.1 | - | 3 | 302 | |||
FQ228844.1 | - | 3 | 302 | |||
Y00697.1 | - | 3 | 295 |