Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNDBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012142.4 | NP_036274.3 | 11 | 1421 | ||
NR_027513.2 | 9 | 1244 | ||||
NR_027514.2 | 10 | 1324 | ||||
NR_045998.1 | 10 | 1341 | ||||
NR_045999.1 | 8 | 1184 | ||||
XM_006720448.1 | XP_006720511.1 | 9 | 1343 | |||
GenBank mRNA | AF082569.1 | - | 11 | 1280 | ||
AF087852.1 | - | 11 | 1266 | |||
AF113535.1 | - | 9 | 1230 | |||
AF132034.1 | - | 11 | 1236 | |||
AF246144.1 | - | 11 | 1238 | |||
AK075146.1 | - | 11 | 1583 | |||
AK075296.1 | - | 9 | 1235 | |||
AK225143.1 | - | 11 | 1236 | |||
AK225645.1 | - | 11 | 1250 | |||
AK290665.1 | - | 11 | 1250 | |||
AK290707.1 | - | 10 | 1203 | |||
AK295017.1 | - | 10 | 1153 | |||
BC009689.1 | - | 11 | 1264 | 3896421 |