Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il2ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013163.1 | NP_037295.1 | 2 | 259 | ||
GenBank mRNA | FQ228242.1 | - | 291 | |||
M55049.1 | - | 2 | 259 |