Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ucp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013167.2 | NP_037299.1 | 7 | 1754 | ||
GenBank mRNA | BC072546.1 | - | 7 | 1754 | 7105743 |