Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013168.2 | NP_037300.2 | 14 | 1463 | ||
XM_006242896.1 | XP_006242958.1 | 14 | 1549 | |||
XM_006242897.1 | XP_006242959.1 | 13 | 1471 | |||
XM_008766148.1 | XP_008764370.1 | 13 | 1633 | |||
XM_008766149.1 | XP_008764371.1 | 13 | 1570 | |||
GenBank mRNA | BC088162.1 | - | 14 | 1463 | 7375228 | |
FQ225972.1 | - | 8 | 948 | |||
FQ226062.1 | - | 14 | 1386 | |||
FQ230962.1 | - | 8 | 948 | |||
FQ231091.1 | - | 14 | 1448 | |||
FQ234793.1 | - | 6 | 835 | |||
FQ234870.1 | - | 12 | 1619 | |||
X06827.1 | - | 14 | 1336 |