Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171713.1 | NP_001165184.1 | 14 | 1254 | ||
NM_012157.3 | NP_036289.3 | 15 | 1461 | |||
XM_005265014.2 | XP_005265071.1 | 15 | 1574 | |||
XM_005265015.3 | XP_005265072.1 | 15 | 1794 | |||
XM_005265016.2 | XP_005265073.1 | 15 | 1538 | |||
XM_005265017.3 | XP_005265074.1 | 15 | 1477 | |||
XM_011533559.2 | XP_011531861.1 | 16 | 1739 | |||
XM_017006092.1 | XP_016861581.1 | 16 | 1631 | |||
XM_017006093.1 | XP_016861582.1 | 15 | 1621 | |||
XM_017006094.1 | XP_016861583.1 | 16 | 1561 | |||
GenBank mRNA | AF176518.1 | - | 15 | 1396 | ||
AK001271.1 | - | 15 | 1461 | |||
AK001438.1 | - | 15 | 1429 | |||
AK096492.1 | - | 2 | 1876 | |||
AK297534.1 | - | 15 | 1445 | |||
AK298967.1 | - | 14 | 1223 | |||
AK303764.1 | - | 13 | 1321 | |||
AL049953.1 | - | 8 | 871 | |||
BC031556.1 | - | 15 | 1405 | 5197151 |