Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013191.1 | NP_037323.1 | 3 | 344 | ||
XM_008772868.1 | XP_008771090.1 | 3 | 343 | |||
GenBank mRNA | BC087026.1 | - | 3 | 282 | 7193740 | |
FQ213381.1 | - | 3 | 286 | |||
FQ213687.1 | - | 3 | 336 | |||
FQ213774.1 | - | 3 | 338 | |||
FQ213840.1 | - | 3 | 294 | |||
FQ214218.1 | - | 3 | 288 | |||
FQ214292.1 | - | 3 | 317 | |||
FQ214554.1 | - | 3 | 286 | |||
M15705.1 | - | 2 | 211 | |||
M54919.1 | - | 3 | 344 | |||
X01090.1 | - | 3 | 344 |