Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXW2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012164.3 | NP_036296.2 | 3 | 621 | ||
XM_005251910.3 | XP_005251967.1 | 3 | 624 | |||
XM_005251913.2 | XP_005251970.1 | 3 | 523 | |||
XM_006717049.3 | XP_006717112.1 | 3 | 806 | |||
XM_006717051.2 | XP_006717114.1 | 3 | 861 | |||
XM_011518518.1 | XP_011516820.1 | 3 | 861 | |||
XM_017014615.1 | XP_016870104.1 | 3 | 624 | |||
XM_017014616.1 | XP_016870105.1 | 3 | 720 | |||
XM_017014617.1 | XP_016870106.1 | 3 | 744 | |||
GenBank mRNA | AF129531.1 | - | 1 | 383 | ||
AF145024.1 | - | 3 | 528 | |||
AF176698.1 | - | 2 | 506 | |||
AF193594.1 | - | 3 | 528 | |||
AK022484.1 | - | 3 | 571 | |||
AK300066.1 | - | 3 | 452 | |||
AK314385.1 | - | 3 | 806 | |||
BC018738.1 | - | 3 | 537 | 4554961 | ||
BC110334.1 | - | 2 | 521 | 5741924 |