Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bglap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013414.1 | NP_038200.1 | 4 | 462 | ||
XM_006232594.2 | XP_006232656.1 | 5 | 3519 | |||
GenBank mRNA | M11777.1 | - | 4 | 442 | ||
X04141.1 | - | 4 | 461 |