Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017125.3 | NP_058821.1 | 6, 7 | 635 | ||
XM_008765024.1 | XP_008763246.1 | 6, 7 | 697 | |||
XM_008765025.1 | XP_008763247.1 | 6, 7 | 900 | |||
GenBank mRNA | AF159104.1 | - | 2, 3 | 259 | ||
BC063173.1 | - | 6, 7 | 638 | 6921594 | ||
FQ214468.1 | - | 6, 7 | 702 | |||
FQ221333.1 | - | 6, 7 | 639 | |||
FQ222038.1 | - | 6, 7 | 618 | |||
FQ230116.1 | - | 6, 7 | 606 | |||
X61654.1 | - | 6, 7 | 635 |