Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017125.3 | NP_058821.1 | 8 | 836 | ||
XM_008765024.1 | XP_008763246.1 | 8 | 898 | |||
XM_008765025.1 | XP_008763247.1 | 8 | 1101 | |||
GenBank mRNA | AF159104.1 | - | 4 | 436 | ||
BC063173.1 | - | 8 | 839 | 6921594 | ||
FQ214468.1 | - | 8 | 903 | |||
FQ221333.1 | - | 8 | 840 | |||
FQ222038.1 | - | 8 | 819 | |||
FQ230116.1 | - | 8 | 807 | |||
X61654.1 | - | 8 | 836 |