Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017127.1 | NP_058823.1 | 4 | 936 | ||
XM_006230707.2 | XP_006230769.1 | 6 | 1959 | |||
XM_006230708.2 | XP_006230770.1 | 5 | 1928 | |||
XM_008760090.1 | XP_008758312.1 | 4 | 3251 | |||
XM_008760091.1 | XP_008758313.1 | 5 | 3411 | |||
GenBank mRNA | BC081821.1 | - | 4 | 912 | 7109261 | |
D10262.1 | - | 4 | 936 | |||
D37885.1 | - | 5 | 692 |