Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017129.1 | NP_058825.1 | 2 | 158 | ||
XM_006230221.2 | XP_006230283.1 | 2 | 921 | |||
GenBank mRNA | BC072690.1 | - | 2 | 147 | 7096800 | |
D78591.1 | - | 2 | 158 |