Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017130.2 | NP_058826.2 | 2, 3 | 389 | ||
XM_006245361.2 | XP_006245423.1 | 2, 3 | 329 | |||
XM_006245362.2 | XP_006245424.1 | 2, 3 | 993 | |||
XM_006245363.2 | XP_006245425.1 | 2, 3 | 340 | |||
XM_006245364.1 | XP_006245426.1 | 2, 3 | 295 | |||
XM_006245365.2 | XP_006245427.1 | 2, 3 | 336 | |||
XM_008767209.1 | XP_008765431.1 | 3, 4 | 521 | |||
XM_008767210.1 | XP_008765432.1 | 3, 4 | 467 | |||
XM_008767211.1 | XP_008765433.1 | 2, 3 | 475 | |||
GenBank mRNA | BC091154.1 | - | 2, 3 | 318 | 7323285 | |
D16300.1 | - | 2, 3 | 389 | |||
FQ213221.1 | - | 2, 3 | 352 | |||
FQ216178.1 | - | 2, 3 | 264 |