Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casq2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017131.2 | NP_058827.3 | 3 | 494 | ||
GenBank mRNA | BC072547.1 | - | 3 | 481 | 7105911 | |
CK602439.1 | - | 3 | 494 | |||
U33287.1 | - | 3 | 475 |