Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017137.1 | NP_058833.1 | 1247 | |||
XM_006248562.2 | XP_006248624.1 | 1740 | ||||
XM_006248563.2 | XP_006248625.1 | 1740 | ||||
GenBank mRNA | X64139.1 | - | 1247 |
Gene Symbol: LOC100912455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751081.3 | XP_003751129.1 | 10 | 1740 | ||
XM_006248554.2 | XP_006248616.1 | 10 | 1740 |