Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017143.2 | NP_058839.1 | 2, 3 | 327 | ||
GenBank mRNA | BC088151.1 | - | 2, 3 | 327 | 7383301 | |
FQ211285.1 | - | 2, 3 | 251 | |||
FQ219425.1 | - | 2, 3 | 245 | |||
X79807.1 | - | 2, 3 | 239 |