Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017145.2 | NP_058841.1 | 5 | 823 | ||
GenBank mRNA | AF063851.1 | - | 5 | 824 | ||
FQ220196.1 | - | 5 | 823 | |||
FQ220424.1 | - | 5 | 785 | |||
FQ220659.1 | - | 5 | 823 | |||
FQ220808.1 | - | 5 | 856 | |||
FQ221313.1 | - | 5 | 823 | |||
FQ223437.1 | - | 2 | 254 | |||
FQ228378.1 | - | 5 | 823 | |||
FQ229265.1 | - | 5 | 823 | |||
FQ229582.1 | - | 5 | 820 | |||
FQ229760.1 | - | 5 | 823 |