Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017151.2 | NP_058847.1 | 3, 4 | 338 | ||
GenBank mRNA | AY390387.1 | - | 2, 3 | 266 | ||
FQ211784.1 | - | 3, 4 | 338 | |||
FQ217117.1 | - | 3, 4 | 339 | |||
FQ217388.1 | - | 3, 4 | 338 | |||
FQ217914.1 | - | 3, 4 | 338 | |||
FQ220999.1 | - | 3, 4 | 338 | |||
FQ221086.1 | - | 3, 4 | 339 | |||
FQ221189.1 | - | 3, 4 | 338 | |||
FQ221217.1 | - | 3, 4 | 339 | |||
FQ221800.1 | - | 3, 4 | 338 | |||
FQ222137.1 | - | 3, 4 | 339 | |||
FQ222200.1 | - | 3, 4 | 338 | |||
FQ222233.1 | - | 3, 4 | 337 | |||
FQ222539.1 | - | 3, 4 | 338 | |||
FQ222610.1 | - | 3, 4 | 338 | |||
FQ222928.1 | - | 3, 4 | 338 | |||
FQ223014.1 | - | 2, 3 | 340 | |||
FQ223247.1 | - | 3, 4 | 338 | |||
FQ223311.1 | - | 3, 4 | 339 | |||
FQ223478.1 | - | 3, 4 | 341 | |||
FQ223811.1 | - | 3, 4 | 338 | |||
FQ223860.1 | - | 3, 4 | 334 | |||
FQ228367.1 | - | 3, 4 | 346 | |||
FQ228405.1 | - | 3, 4 | 341 | |||
M19393.1 | - | 3, 4 | 338 |