Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017152.3 | NP_058848.2 | 2 | 169 | ||
GenBank mRNA | BC058484.1 | - | 2 | 173 | 6921184 | |
FQ209589.1 | - | 2 | 166 | |||
FQ210096.1 | - | 2 | 159 | |||
FQ221123.1 | - | 2 | 164 | |||
FQ221680.1 | - | 2 | 159 | |||
FQ222308.1 | - | 2 | 159 | |||
FQ222382.1 | - | 2 | 171 | |||
FQ222858.1 | - | 2 | 164 | |||
FQ223398.1 | - | 2 | 166 | |||
FQ224175.1 | - | 2 | 159 | |||
FQ228331.1 | - | 2 | 164 | |||
FQ228517.1 | - | 2 | 166 | |||
FQ229910.1 | - | 2 | 166 | |||
K02933.1 | - | 2 | 167 |
Gene Symbol: LOC100365810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006243604.2 | XP_006243666.1 | 1 | 165 |