Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adora1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017155.2 | NP_058851.2 | 2 | 515 | ||
XM_006249859.2 | XP_006249921.1 | 3 | 806 | |||
XM_006249860.2 | XP_006249922.1 | 3 | 581 | |||
XM_006249861.2 | XP_006249923.1 | 3 | 745 | |||
XM_008769552.1 | XP_008767774.1 | 3 | 968 | |||
XM_008769553.1 | XP_008767775.1 | 3 | 907 | |||
GenBank mRNA | AB001089.1 | - | 2 | 452 | ||
AF042079.1 | - | 2 | 483 | |||
M69045.1 | - | 2 | 515 | |||
Y12519.1 | - | 2 | 867 |
Set Membership: |
G-protein coupled receptor |