Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017159.1 | NP_058855.1 | 10 | 886 | ||
GenBank mRNA | BC089809.1 | - | 10 | 915 | 7325816 | |
M58308.1 | - | 10 | 886 |