Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAVCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173393.2 | NP_001166864.1 | 8 | 1188 | ||
NM_012206.3 | NP_036338.2 | 7 | 1275 | |||
GenBank mRNA | AF043724.1 | - | 8 | 1013 | ||
BC013325.1 | - | 7 | 1145 | 3826378 | ||
CR457114.1 | - | 7 | 977 | |||
HQ412639.1 | - | 7 | 962 | |||
HQ412640.1 | - | 7 | 962 | |||
JX049978.1 | - | 7 | 962 |