Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017169.1 | NP_058865.1 | 6 | 861 | ||
XM_006255244.1 | XP_006255306.1 | 6 | 1042 | |||
GenBank mRNA | BC058481.1 | - | 6 | 888 | 6921522 | |
FQ211079.1 | - | 6 | 877 | |||
FQ215547.1 | - | 6 | 913 | |||
FQ221561.1 | - | 6 | 882 | |||
FQ225602.1 | - | 6 | 884 | |||
FQ227580.1 | - | 6 | 875 | |||
FQ233937.1 | - | 6 | 884 | |||
FQ234358.1 | - | 6 | 913 | |||
FQ234566.1 | - | 6 | 979 | |||
U06099.1 | - | 6 | 861 |