Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004439.1 | NP_001004439.1 | 27 | 3329 | ||
XM_005254228.3 | XP_005254285.1 | 25 | 3551 | |||
XM_011521363.2 | XP_011519665.1 | 25 | 3281 | |||
GenBank mRNA | AF109681.1 | - | 27 | 3317 | ||
AF111799.1 | - | 19 | 2294 | |||
AF137378.2 | - | 27 | 3332 | |||
AK292445.1 | - | 27 | 3338 | |||
AL359064.1 | - | 5 | 385 |