Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100909648 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289800.1 | NP_001276729.1 | 1 | 474 | ||
XM_008769404.1 | XP_008767626.1 | 1 | 659 |
Gene Symbol: Avpr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017205.3 | NP_058901.3 | 2 | 1006 | ||
XM_008769445.1 | XP_008767667.1 | 1 | 666 | |||
GenBank mRNA | D45400.1 | - | 1 | 523 | ||
U27322.1 | - | 1 | 899 |
Set Membership: |
G-protein coupled receptor |