Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017212.2 | NP_058908.2 | 10 | 3198 | ||
XM_008768268.1 | XP_008766490.1 | 14 | 4371 | |||
XM_008768269.1 | XP_008766491.1 | 13 | 4284 | |||
XM_008768270.1 | XP_008766492.1 | 13 | 4279 | |||
XM_008768271.1 | XP_008766493.1 | 12 | 4197 | |||
XM_008768272.1 | XP_008766494.1 | 13 | 4172 | |||
XM_008768273.1 | XP_008766495.1 | 12 | 4079 | |||
XM_008768274.1 | XP_008766496.1 | 13 | 3605 | |||
XM_008768275.1 | XP_008766497.1 | 12 | 3518 | |||
XM_008768277.1 | XP_008766499.1 | 12 | 3404 | |||
XM_008768278.1 | XP_008766500.1 | 11 | 3317 | |||
XM_008768279.1 | XP_008766501.1 | 11 | 3312 | |||
XM_008768280.1 | XP_008766502.1 | 10 | 3224 | |||
XM_008768281.1 | XP_008766503.1 | 9 | 3137 | |||
XM_008768282.1 | XP_008766504.1 | 15 | 4426 | |||
GenBank mRNA | CO394568.1 | - | 1 | 371 | ||
X79321.1 | - | 10 | 3276 |