Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitpna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017231.1 | NP_058927.1 | 12 | 1603 | ||
GenBank mRNA | M25758.1 | - | 12 | 1603 |