Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psme2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017257.2 | NP_058953.1 | 3 | 122 | ||
GenBank mRNA | BC058486.1 | - | 3 | 160 | 6918985 | |
BC097331.1 | - | 3 | 122 | 7389180 | ||
D45250.1 | - | 3 | 172 | |||
FQ226805.1 | - | 3 | 153 | |||
FQ227049.1 | - | 3 | 166 | |||
FQ232840.1 | - | 3 | 153 | |||
FQ233763.1 | - | 3 | 148 |