Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017259.1 | NP_058955.1 | 1, 2 | 204 | ||
GenBank mRNA | BC072493.1 | - | 1, 2 | 163 | 7097327 | |
M60921.1 | - | 1, 2 | 204 |