Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gria2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083811.1 | NP_001077280.1 | 3 | 841 | ||
NM_017261.2 | NP_058957.1 | 3 | 841 | |||
GenBank mRNA | AF164344.1 | - | 3 | 760 | ||
M36419.1 | - | 3 | 411 | |||
M38061.1 | - | 3 | 841 | |||
M85035.1 | - | 3 | 726 | |||
X54655.1 | - | 3 | 677 |