Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgcs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017268.1 | NP_058964.1 | 6 | 954 | ||
XM_006231959.1 | XP_006232021.1 | 5 | 1123 | |||
GenBank mRNA | AY724522.1 | - | 5 | 940 | ||
FQ211444.1 | - | 5 | 926 | |||
X52625.1 | - | 6 | 954 |