Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317960.1 | NP_001304889.1 | 23 | 2441 | ||
NM_012255.4 | NP_036387.2 | 23 | 2281 | |||
XM_017027723.1 | XP_016883212.1 | 23 | 2234 | |||
GenBank mRNA | AF064257.1 | - | 23 | 2234 | ||
AF152169.1 | - | 23 | 2184 | |||
AK000084.1 | - | 12 | 1096 | |||
AK001573.1 | - | 23 | 2251 | |||
AK302846.1 | - | 23 | 2145 | |||
AK303312.1 | - | 22 | 2092 | |||
AK314602.1 | - | 23 | 2220 | |||
AL136841.1 | - | 23 | 2207 | |||
AY382900.1 | - | 22 | 2096 | |||
BC006417.1 | - | 11 | 1056 |