Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017275.1 | NP_058971.1 | 8 | 905 | ||
XM_006229572.2 | XP_006229634.1 | 8 | 737 | |||
XM_006229573.2 | XP_006229635.1 | 8 | 761 | |||
XM_006229574.2 | XP_006229636.1 | 8 | 711 | |||
XM_006229575.2 | XP_006229637.1 | 8 | 715 | |||
XM_008773628.1 | XP_008771850.1 | 8 | 830 | |||
XM_008773629.1 | XP_008771851.1 | 7 | 708 | |||
GenBank mRNA | AB004267.1 | - | 8 | 905 | ||
D86556.1 | - | 8 | 756 |