Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017283.3 | NP_058979.1 | 2 | 168 | ||
XM_006240098.2 | XP_006240160.1 | 2 | 126 | |||
GenBank mRNA | BC062232.1 | - | 2 | 169 | 6888689 | |
D10755.1 | - | 2 | 157 | |||
FQ212808.1 | - | 270 | ||||
FQ217234.1 | - | 2 | 144 | |||
FQ220193.1 | - | 2 | 195 |
Gene Symbol: LOC100125377 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC089066.1 | - | 354 | 7319366 |