Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110139.2 | NP_001103609.1 | 3, 4 | 719 | ||
NM_001110823.2 | NP_001104293.1 | 3, 4 | 719 | |||
GenBank mRNA | J04022.1 | - | 3, 4 | 717 | ||
J04023.1 | - | 3, 4 | 352 | |||
J04024.1 | - | 3, 4 | 451 | |||
X15635.1 | - | 3, 4 | 420 |