Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110139.2 | NP_001103609.1 | 7 | 1109 | ||
NM_001110823.2 | NP_001104293.1 | 7 | 1109 | |||
GenBank mRNA | AF043106.1 | - | 2 | 81 | ||
CK356454.1 | - | 1 | 68 | |||
J04022.1 | - | 7 | 1107 | |||
J04023.1 | - | 7 | 742 | |||
J04024.1 | - | 7 | 841 | |||
X15635.1 | - | 7 | 810 |