Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017296.1 | NP_058992.1 | 1 | 142 | ||
XM_006247567.2 | XP_006247629.1 | 2 | 3475 | |||
GenBank mRNA | AF021137.1 | - | 1 | 142 | ||
L48490.1 | - | 1 | 142 |