Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035232.1 | NP_001030309.1 | 2 | 286 | ||
NM_017299.2 | NP_058995.2 | 2 | 419 | |||
XM_006256295.2 | XP_006256357.1 | 2 | 1308 | |||
XM_006256296.2 | XP_006256358.1 | 2 | 311 | |||
XM_006256297.2 | XP_006256359.1 | 2 | 735 | |||
XM_006256298.2 | XP_006256360.1 | 2 | 297 | |||
GenBank mRNA | AF099009.1 | - | 2 | 428 | ||
AF099010.1 | - | 2 | 428 | |||
AF173642.1 | - | 2 | 265 | |||
AY756271.1 | - | 2 | 265 | |||
AY756272.1 | - | 2 | 265 | |||
AY756273.1 | - | 2 | 265 | |||
BC085742.1 | - | 2 | 286 | 7193913 | ||
U38180.1 | - | 2 | 411 |