Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM184B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195071.1 | NP_001182000.1 | 3, 4 | 589 | ||
NM_001195072.1 | NP_001182001.1 | 4, 5 | 675 | |||
NM_012264.4 | NP_036396.2 | 3, 4 | 574 | |||
XM_011530112.2 | XP_011528414.2 | 3, 4 | 623 | |||
XM_011530113.1 | XP_011528415.1 | 3, 4 | 553 | |||
XM_011530114.1 | XP_011528416.1 | 3, 4 | 585 | |||
XM_011530116.2 | XP_011528418.2 | 3, 4 | 615 | |||
XM_011530117.1 | XP_011528419.1 | 4, 5 | 651 | |||
XM_011530118.1 | XP_011528420.1 | 4, 5 | 625 | |||
XM_017028754.1 | XP_016884243.1 | 3, 4 | 624 | |||
XM_017028755.1 | XP_016884244.1 | 3, 4 | 617 | |||
XM_017028756.1 | XP_016884245.1 | 3, 4 | 553 | |||
XM_017028758.1 | XP_016884247.1 | 3, 4 | 553 | |||
GenBank mRNA | AB097053.1 | - | 3, 4 | 545 | ||
AK075417.1 | - | 1, 2 | 62 | |||
AK292652.1 | - | 3, 4 | 574 | |||
AL096879.1 | - | 2, 3 | 267 | |||
BC015489.1 | - | 3, 4 | 530 | 3896682 | ||
BI197128.1 | - | 2, 3 | 268 | |||
BX648345.1 | - | 3, 4 | 625 | |||
CR456413.1 | - | 2, 3 | 408 | |||
CR749859.1 | - | 4, 5 | 581 |