Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135004.2 | NP_001128476.2 | 3 | 504 | ||
NM_001135005.2 | NP_001128477.1 | 2 | 711 | |||
NM_012266.5 | NP_036398.3 | 2 | 359 | |||
XM_005251428.3 | XP_005251485.1 | 2 | 278 | |||
XM_006716751.2 | XP_006716814.1 | 3 | 368 | |||
XM_011517842.2 | XP_011516144.1 | 3 | 945 | |||
GenBank mRNA | AA351558.1 | - | 3 | 361 | ||
AF087870.1 | - | 3 | 361 | |||
AF088982.1 | - | 2 | 262 | |||
AK023253.1 | - | 2 | 255 | |||
AK299647.1 | - | 2 | 711 | |||
AK314671.1 | - | 2 | 236 | |||
AL536638.3 | - | 3 | 392 | |||
BI915181.1 | - | 2 | 663 | |||
BU845478.1 | - | 2 | 272 | |||
BX419166.2 | - | 2 | 700 |