Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017319.1 | NP_059015.1 | 12 | 1464 | ||
GenBank mRNA | BC062393.1 | - | 12 | 1501 | 5621329 | |
D63378.1 | - | 12 | 1464 | |||
FQ211627.1 | - | 1563 | ||||
X12355.1 | - | 12 | 1457 |