Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019140.2 | NP_062013.1 | 2, 3 | 285 | ||
GenBank mRNA | BC105753.1 | - | 528 | 5599711 | ||
L12329.1 | - | 2, 3 | 285 | |||
L19180.1 | - | 131 | ||||
L19181.1 | - | 1, 2 | 131 | |||
L19933.1 | - | 1, 2 | 199 |