Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012287.5 | NP_036419.3 | 2 | 325 | ||
XM_006713557.3 | XP_006713620.1 | 2 | 325 | |||
XM_011512602.2 | XP_011510904.1 | 2 | 325 | |||
XM_011512603.2 | XP_011510905.1 | 2 | 325 | |||
XM_011512604.2 | XP_011510906.1 | 3 | 484 | |||
XM_011512605.2 | XP_011510907.1 | 2 | 246 | |||
XM_011512606.2 | XP_011510908.1 | 2 | 227 | |||
XM_017006046.1 | XP_016861535.1 | 2 | 349 | |||
XM_017006047.1 | XP_016861536.1 | 2 | 349 | |||
XM_017006048.1 | XP_016861537.1 | 2 | 349 | |||
XM_017006049.1 | XP_016861538.1 | 2 | 349 | |||
GenBank mRNA | AJ238248.1 | - | 2 | 144 | ||
AK222635.1 | - | 2 | 280 | |||
AK290369.1 | - | 2 | 257 | |||
BC060767.1 | - | 2 | 260 | 30343546 | ||
D26069.3 | - | 2 | 94 | |||
DA774230.1 | - | 2 | 280 | |||
DB087306.1 | - | 2 | 325 |