Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uts2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019160.2 | NP_062033.1 | 1 | 115 | ||
GenBank mRNA | AF172174.1 | - | 1 | 115 |