Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019165.1 | NP_062038.1 | 5 | 601 | ||
GenBank mRNA | AJ222813.1 | - | 5 | 601 | ||
AY258448.1 | - | 5 | 601 | |||
U77776.1 | - | 5 | 601 | |||
U77777.1 | - | 5 | 544 |