Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012291.4 | NP_036423.4 | 4 | 1296 | ||
XM_006719705.3 | XP_006719768.1 | 4 | 1363 | |||
XM_011539024.2 | XP_011537326.1 | 4 | 1324 | |||
XM_011539025.2 | XP_011537327.1 | 4 | 1335 | |||
XM_017020253.1 | XP_016875742.1 | 4 | 1414 | |||
GenBank mRNA | AY455930.1 | - | 3 | 1205 | ||
BC047603.1 | - | 2 | 94 | |||
D79987.1 | - | 4 | 1343 |